Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007531.hg.1 |
ENST00000534243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007531.hg.1 |
ENST00000534243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007531.hg.1 |
ENST00000534243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007531.hg.1 |
ENST00000534243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010821.hg.1 |
ENST00000532369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010821.hg.1 |
ENST00000532369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010821.hg.1 |
ENST00000532369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010821.hg.1 |
ENST00000532369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010830.hg.1 |
ENST00000533973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010830.hg.1 |
ENST00000533973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010830.hg.1 |
ENST00000533973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010830.hg.1 |
ENST00000533973.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007580.hg.1 |
ENST00000533055.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010100.hg.1 |
ENST00000508690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010100.hg.1 |
ENST00000508690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007530.hg.1 |
ENST00000525520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007530.hg.1 |
ENST00000525520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007530.hg.1 |
ENST00000525520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007549.hg.1 |
ENST00000532682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007549.hg.1 |
ENST00000532682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007549.hg.1 |
ENST00000532682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007549.hg.1 |
ENST00000532682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007549.hg.1 |
ENST00000532682.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010801.hg.1 |
ENST00000532033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010801.hg.1 |
ENST00000532033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010801.hg.1 |
ENST00000532033.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010801.hg.1 |
ENST00000532033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007530.hg.1 |
ENST00000525520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007530.hg.1 |
ENST00000525520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007530.hg.1 |
ENST00000525520.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010830.hg.1 |
ENST00000533973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010830.hg.1 |
ENST00000533973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010830.hg.1 |
ENST00000533973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010830.hg.1 |
ENST00000533973.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007550.hg.1 |
ENST00000534129.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010100.hg.1 |
ENST00000508690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010100.hg.1 |
ENST00000508690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010821.hg.1 |
ENST00000532369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010821.hg.1 |
ENST00000532369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010821.hg.1 |
ENST00000532369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010821.hg.1 |
ENST00000532369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007580.hg.1 |
ENST00000533055.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007549.hg.1 |
ENST00000532682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007549.hg.1 |
ENST00000532682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007549.hg.1 |
ENST00000532682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007549.hg.1 |
ENST00000532682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007549.hg.1 |
ENST00000532682.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007531.hg.1 |
ENST00000534243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007531.hg.1 |
ENST00000534243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007531.hg.1 |
ENST00000534243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007531.hg.1 |
ENST00000534243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010801.hg.1 |
ENST00000532033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010801.hg.1 |
ENST00000532033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010801.hg.1 |
ENST00000532033.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010801.hg.1 |
ENST00000532033.1 |
Ensembl |