Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000572145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000572145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000572145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000572145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000572145.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000574295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000574295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000574295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
ENST00000574295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000300574.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000398970.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000572145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000572145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000572145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000572145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000572145.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000574295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000574295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000574295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
ENST00000574295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_005206.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009372.hg.1 |
NM_016823.3 |
Refseq |