Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
kazrin, periplakin interacting protein |
KAZN | KAZ | Sense | Chr. 1 - 13893387 - 15118048 on NCBI Build GRCh38 | |
Downstream | TMEM51 antisense RNA 1 | TMEM51-AS1 | C1orf126 | 783.0kb | Anti-sense | Chr. 1 - 15111815 - 15152464 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |