Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | G-patch domain containing 2 | GPATCH2 |
CT110, GPATC2, PPP1R30 |
Sense | Chr. 1 - 217426992 - 217631312 on NCBI Build GRCh38 | |
Downstream | spermatogenesis associated 17 | SPATA17 |
IQCH, MSRG-11, MSRG11 |
174.0kb | Anti-sense | Chr. 1 - 217631324 - 217868151 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |