Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSU72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK129493.1 | - | 1 | 651 | 62 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BM671318.1 | - | 1 | Chr. 1 - 1560846 - 1561531 [+] on Build GRCh38 | 62 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 240 | TMEM240 |
C1orf70, SCA21 |
-20.7kb | Sense | Chr. 1 - 1534778 - 1540360 on NCBI Build GRCh38 |
Downstream | fibronectin type III domain containing 10 | FNDC10 | C1orf233 | 35.9kb | Sense | Chr. 1 - 1598008 - 1600096 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |