Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | STAG3L5P-PVRIG2P-PILRB readthrough | STAG3L5P-PVRIG2P-PILRB | LOC101735302-LOC101752334-PILRB | -7.2kb | Sense | Chr. 7 - 100336065 - 100367831 on NCBI Build GRCh38 |
Downstream | zinc finger CW-type and PWWP domain containing 1 | ZCWPW1 | ZCW1 | 25.0kb | Anti-sense | Chr. 7 - 100400826 - 100429239 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |