Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000413821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000413821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000413821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000423427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000423427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000423427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000469564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000469564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000469564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000469564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000469564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000469727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000469727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000469727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000469727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000469727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000482120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000482120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000490929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000490929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
ENST00000490929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000361589.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000401917.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000413821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000413821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000413821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000415045.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000423427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000423427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000423427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000457714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000466350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000469564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000469564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000469564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000469564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000469564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000469727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000469727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000469727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000469727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000469727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000482120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000482120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000490929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000490929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
ENST00000490929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009531.hg.1 |
NM_014932.3 |
Refseq |