Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX640965.1 | - | 1 | 647 | 72 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 22 member 2 | SLC22A2 | OCT2 | -187.7kb | Anti-sense | Chr. 6 - 160216762 - 160258931 on NCBI Build GRCh38 |
Downstream |
lipoprotein(a) like 2, pseudogene |
LPAL2 |
APOA2, APOAL, APOARGC, apo(a)rg-C |
14.0kb | Anti-sense | Chr. 6 - 160466555 - 160511124 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |