Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | leucine rich repeat containing 57 | LRRC57 | Sense | Chr. 15 - 42528332 - 42548804 on NCBI Build GRCh38 | ||
Downstream | HAUS augmin like complex subunit 2 | HAUS2 |
C15orf25, CEP27, HsT17025 |
9.4kb | Anti-sense | Chr. 15 - 42548813 - 42569994 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |