Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101929125 | LOC101929125 | Anti-sense | Chr. 20 - 3808494 - 3812434 on NCBI Build GRCh38 | ||
Downstream | adaptor related protein complex 5 sigma 1 subunit | AP5S1 | C20orf29 | 8.8kb | Anti-sense | Chr. 20 - 3820524 - 3825307 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |