Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | StAR related lipid transfer domain containing 9 | STARD9 | KIF16A | Anti-sense | Chr. 15 - 42575646 - 42720998 on NCBI Build GRCh38 | |
Downstream | codanin 1 | CDAN1 |
CDA1, CDAI, CDAN1A, DLT, PRO1295 |
3.9kb | Sense | Chr. 15 - 42723557 - 42737219 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |