Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | antizyme inhibitor 2 | AZIN2 |
ADC, AZI2, AZIB1, ODC-p, ODC1L, ODCp |
Sense | Chr. 1 - 33081110 - 33162288 on NCBI Build GRCh38 | |
Downstream | zinc finger protein 362 | ZNF362 |
RN, lin-29 |
90.0kb | Sense | Chr. 1 - 33256307 - 33300719 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |