Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | WD repeat domain 20 | WDR20 | DMR | Anti-sense | Chr. 14 - 102139518 - 102224686 on NCBI Build GRCh38 | |
Downstream | MOK protein kinase | MOK |
RAGE, RAGE-1, RAGE1, STK30 |
6.7kb | Sense | Chr. 14 - 102224450 - 102305200 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |