Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ankyrin repeat and SOCS box containing 13 | ASB13 | -14.2kb | Sense | Chr. 10 - 5638857 - 5666595 on NCBI Build GRCh38 | |
Downstream | GDP dissociation inhibitor 2 | GDI2 |
HEL-S-46e, RABGDIB |
79.5kb | Sense | Chr. 10 - 5765222 - 5813549 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |