Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928438 | LOC101928438 | Sense | Chr. 9 - 99585786 - 99819889 on NCBI Build GRCh38 | ||
Downstream | nuclear receptor subfamily 4 group A member 3 | NR4A3 |
CHN, CSMF, MINOR, NOR1, TEC |
16.8kb | Anti-sense | Chr. 9 - 99821855 - 99866893 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |