Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | anthrax toxin receptor 2 | ANTXR2 |
CMG-2, CMG2, HFS, ISH, JHF |
Anti-sense | Chr. 4 - 79901617 - 80073472 on NCBI Build GRCh38 | |
Downstream | PR domain 8 | PRDM8 |
EPM10, PFM5 |
232.6kb | Sense | Chr. 4 - 80183418 - 80204329 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |