Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | FAM13A antisense RNA 1 | FAM13A-AS1 |
FAM13A1OS, FAM13AOS, NCRNA00039 |
-34.1kb | Anti-sense | Chr. 4 - 88709789 - 88730103 on NCBI Build GRCh38 |
Downstream | tigger transposable element derived 2 | TIGD2 | HEL106 | 346.2kb | Anti-sense | Chr. 4 - 89112817 - 89114901 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |