Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
protease, serine 42 |
PRSS42 | TESSP2 | -21.7kb | Sense | Chr. 3 - 46830404 - 46834095 on NCBI Build GRCh38 |
Downstream | myosin light chain 3 | MYL3 |
CMH8, MLC-lV/sb, MLC1SB, MLC1V, VLC1, VLCl |
369.0 | Sense | Chr. 3 - 46857867 - 46863483 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |