Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004975.3 | NP_004966.1 | 2 | 11618 | 76 | |
XM_006723784.3 | XP_006723847.1 | 3 | 11712 | 76 | ||
XM_011528799.2 | XP_011527101.1 | 3 | 11741 | 76 | ||
GenBank mRNA | AK126976.1 | - | 1 | 3651 | 76 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ZNFX1 antisense RNA 1 | ZFAS1 |
C20orf199, HSUP1, HSUP2, NCRNA00275, ZNFX1-AS1 |
-74.6kb | Anti-sense | Chr. 20 - 49278178 - 49289260 on NCBI Build GRCh38 |
Downstream | prostaglandin I2 (prostacyclin) synthase | PTGIS |
CYP8, CYP8A1, PGIS, PTGI |
136.1kb | Sense | Chr. 20 - 49503874 - 49568170 on NCBI Build GRCh38 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |