Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil domain containing 178 | CCDC178 | C18orf34 | -206.1kb | Anti-sense | Chr. 18 - 32937402 - 33441040 on NCBI Build GRCh38 |
Downstream | nucleolar protein 4 | NOL4 |
CT125, HRIHFB2255, NOLP |
200.5kb | Anti-sense | Chr. 18 - 33851100 - 34224834 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |