Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PAP associated domain containing 7 | PAPD7 |
LAK-1, LAK1, POLK, POLS, TRF4, TRF4-1, TRF41, TUTASE5 |
-10.2kb | Anti-sense | Chr. 5 - 6713952 - 6757048 on NCBI Build GRCh38 |
Downstream | microRNA 4278 | MIR4278 | 57.2kb | Sense | Chr. 5 - 6827853 - 6827921 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |