Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011714.hg.1 |
ENST00000517054.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006502.hg.1 |
ENST00000410629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009420.hg.1 |
ENST00000410365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008860.hg.1 |
ENST00000363347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008860.hg.1 |
ENST00000363347.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006576.hg.1 |
ENST00000364312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011190.hg.1 |
ENST00000364072.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010177.hg.1 |
ENST00000384449.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007573.hg.1 |
ENST00000365488.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006957.hg.1 |
ENST00000411008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007845.hg.1 |
ENST00000364709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007042.hg.1 |
ENST00000458780.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007105.hg.1 |
ENST00000516666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011015.hg.1 |
ENST00000384424.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011594.hg.1 |
ENST00000362636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006502.hg.1 |
ENST00000410629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011015.hg.1 |
ENST00000384424.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011714.hg.1 |
ENST00000517054.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006576.hg.1 |
ENST00000364312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011594.hg.1 |
ENST00000362636.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011190.hg.1 |
ENST00000364072.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010177.hg.1 |
ENST00000384449.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009420.hg.1 |
ENST00000410365.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006957.hg.1 |
ENST00000411008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007845.hg.1 |
ENST00000364709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007042.hg.1 |
ENST00000458780.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007573.hg.1 |
ENST00000365488.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008860.hg.1 |
ENST00000363347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008860.hg.1 |
ENST00000363347.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007105.hg.1 |
ENST00000516666.1 |
Ensembl |