Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hepatoma-derived growth factor-related protein 2 | HDGFRP2 |
HDGF-2, HDGF2, HRP-2 |
Sense | Chr. 19 - 4472196 - 4502220 on NCBI Build GRCh38 | |
Downstream | perilipin 4 | PLIN4 |
KIAA1881, S3-12 |
22.6kb | Anti-sense | Chr. 19 - 4502180 - 4520285 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |