Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
small nucleolar RNA, C/D box 56B |
SNORD56B | RNU56B | -72.5kb | Sense | Chr. 14 - 71398337 - 71398407 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC145474 | LOC145474 | 15.5kb | Sense | Chr. 14 - 71487861 - 71489714 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |