Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000506257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000506257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000506257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
ENST00000581282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000322285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000506257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000506257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000506257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
ENST00000581282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_001297716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007820.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153483.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153483.hg.1 |
ENST00000581282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153483.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153483.hg.1 |
ENST00000502798.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153483.hg.1 |
ENST00000581282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153483.hg.1 |
NM_014979.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153483.hg.1 |
TC0500007820.hg.1 |
Clariom D TC |