Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nucleolin | NCL | C23 | -40.0kb | Sense | Chr. 2 - 231454748 - 231464494 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 471 | LINC00471 | C2orf52 | 3.1kb | Sense | Chr. 2 - 231508426 - 231514339 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |