Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | interleukin 12 receptor subunit beta 2 | IL12RB2 | -2.5kb | Sense | Chr. 1 - 67307351 - 67397730 on NCBI Build GRCh38 | |
Downstream | SERPINE1 mRNA binding protein 1 | SERBP1 |
CGI-55, CHD3IP, HABP4L, PAI-RBP1, PAIRBP1 |
7.2kb | Anti-sense | Chr. 1 - 67407810 - 67430440 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |