Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005253351.3 | XP_005253408.1 | 4 | 17004 | 65 | |
XM_011520628.2 | XP_011518930.1 | 14 | 20070 | 65 | ||
XM_011520629.2 | XP_011518931.1 | 15 | 19877 | 65 | ||
XM_017019219.1 | XP_016874708.1 | 14 | 19625 | 65 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1559 | LINC01559 | C12orf36 | -171.3kb | Sense | Chr. 12 - 13370671 - 13376745 on NCBI Build GRCh38 |
Downstream | activating transcription factor 7 interacting protein | ATF7IP |
AM, ATF-IP, MCAF, MCAF1, p621 |
816.9kb | Anti-sense | Chr. 12 - 14365632 - 14502935 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |