Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | methyltransferase like 25 | METTL25 | C12orf26 | -33.4kb | Sense | Chr. 12 - 82358497 - 82479239 on NCBI Build GRCh38 |
Downstream | transmembrane and tetratricopeptide repeat containing 2 | TMTC2 | IBDBP1 | 171.1kb | Sense | Chr. 12 - 82686952 - 83134868 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |