Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Ral GTPase activating protein catalytic alpha subunit 2 | RALGAPA2 |
AS250, C20orf74, bA287B20.1, dJ1049G11, dJ1049G11.4, p220 |
-435.1kb | Anti-sense | Chr. 20 - 20389558 - 20712638 on NCBI Build GRCh38 |
Downstream | KIZ antisense RNA 1 | KIZ-AS1 | 40.2kb | Anti-sense | Chr. 20 - 21194404 - 21218289 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |