Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | GLIS family zinc finger 3 | GLIS3 |
NDH, ZNF515 |
-181.3kb | Anti-sense | Chr. 9 - 3824127 - 4310694 on NCBI Build GRCh38 |
Downstream | spermatogenesis associated 6 like | SPATA6L |
C9orf68, bA6J24.2 |
103.0kb | Anti-sense | Chr. 9 - 4540092 - 4666674 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |