Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 138-1 | MIR138-1 |
MIRN138-1, mir-138-1 |
-1.5kb | Anti-sense | Chr. 3 - 44114212 - 44114310 on NCBI Build GRCh38 |
Downstream | testis and ovary specific PAZ domain containing 1 | TOPAZ1 | C3orf77 | 124.4kb | Anti-sense | Chr. 3 - 44241886 - 44337071 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |