Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000147.4 | NP_000138.2 | 3 - 4 | 747 | 69 | |
XM_005245821.2 | XP_005245878.1 | 3 - 4 | 884 | 69 | ||
XM_011541167.2 | XP_011539469.1 | 3 - 4 | 332 | 69 | ||
XM_017000905.1 | XP_016856394.1 | 3 - 4 | 429 | 69 | ||
GenBank mRNA | AB074175.1 | - | 3 - 4 | 668 | 69 | |
AK092914.1 | - | 4 - 5 | 602 | 69 | ||
AK095443.1 | - | 4 - 5 | 619 | 69 | ||
AK095455.1 | - | 4 - 5 | 633 | 69 | ||
AK225468.1 | - | 3 - 4 | 668 | 69 | ||
AK295535.1 | - | 3 - 4 | 851 | 69 | ||
AK301302.1 | - | 5 - 6 | 597 | 69 | ||
AK310143.1 | - | 4 - 5 | 730 | 69 | ||
AK314649.1 | - | 3 - 4 | 668 | 69 | ||
BC017338.2 | - | 3 - 4 | 670 | 4871788 | 69 | |
M10355.1 | - | 3 - 4 | 442 | 69 | ||
M29877.1 | - | 3 - 4 | 667 | 69 | ||
X01390.1 | - | 3 - 4 | 442 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons |