Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173393.2 | NP_001166864.1 | 3 - 4 | 588 | 85 | |
NM_001308156.1 | NP_001295085.1 | 3 - 4 | 709 | 85 | ||
NM_012206.3 | NP_036338.2 | 2 - 3 | 675 | 85 | ||
XM_017009339.1 | XP_016864828.1 | 2 - 3 | 389 | 85 | ||
XM_017009340.1 | XP_016864829.1 | 2 - 3 | 389 | 85 | ||
GenBank mRNA | AF043724.1 | - | 3 - 4 | 428 | 85 | |
AK298259.1 | - | 3 - 4 | 709 | 85 | ||
BC013325.1 | - | 2 - 3 | 545 | 3826378 | 85 | |
CR457114.1 | - | 2 - 3 | 377 | 85 | ||
DA384612.1 | - | 3 - 4 | 432 | 85 | ||
HQ412639.1 | - | 2 - 3 | 377 | 85 | ||
HQ412640.1 | - | 2 - 3 | 377 | 85 | ||
HQ412641.1 | - | 2 - 3 | 377 | 85 | ||
JX049978.1 | - | 2 - 3 | 377 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |