Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024941.hg.1 |
TC19002615.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024941.hg.1 |
ENST00000251390.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024941.hg.1 |
ENST00000391745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024941.hg.1 |
NM_006865.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024943.hg.1 |
TC19002615.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024943.hg.1 |
ENST00000251390.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024943.hg.1 |
ENST00000391744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024943.hg.1 |
ENST00000391745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024943.hg.1 |
NM_001172654.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024943.hg.1 |
NM_006865.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024939.hg.1 |
TC19002615.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024939.hg.1 |
ENST00000251390.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024939.hg.1 |
ENST00000391744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024939.hg.1 |
ENST00000391745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024939.hg.1 |
NM_001172654.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024939.hg.1 |
NM_006865.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024946.hg.1 |
TC19002615.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024946.hg.1 |
ENST00000251390.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024946.hg.1 |
ENST00000391745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024946.hg.1 |
NM_001172654.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024946.hg.1 |
NM_006865.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024948.hg.1 |
TC19002615.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024948.hg.1 |
ENST00000251390.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024948.hg.1 |
NM_006865.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024942.hg.1 |
TC19002615.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024942.hg.1 |
ENST00000251390.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024942.hg.1 |
ENST00000391744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024942.hg.1 |
ENST00000391745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024942.hg.1 |
NM_001172654.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024942.hg.1 |
NM_006865.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024940.hg.1 |
TC19002615.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024940.hg.1 |
ENST00000251390.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024940.hg.1 |
ENST00000391744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024940.hg.1 |
ENST00000391745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024940.hg.1 |
NM_001172654.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024940.hg.1 |
NM_006865.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024944.hg.1 |
TC19002615.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024944.hg.1 |
ENST00000251390.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024944.hg.1 |
ENST00000391744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024944.hg.1 |
ENST00000391745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024944.hg.1 |
NM_001172654.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024944.hg.1 |
NM_006865.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024945.hg.1 |
TC19002615.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024945.hg.1 |
ENST00000251390.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024945.hg.1 |
ENST00000391744.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024945.hg.1 |
ENST00000391745.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024945.hg.1 |
NM_001172654.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024945.hg.1 |
NM_006865.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011802.hg.1 |
ENST00000444199.1 |
Ensembl |