Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256672.1 | NP_001243601.1 | 6 | 1060 | 53 | |
NM_001256673.1 | NP_001243602.1 | 5 | 913 | 53 | ||
NM_001256674.1 | NP_001243603.1 | 5 | 910 | 53 | ||
NM_001256676.1 | NP_001243605.1 | 6 | 874 | 53 | ||
NM_001256677.1 | NP_001243606.1 | 7 | 1191 | 53 | ||
NM_001324226.1 | NP_001311155.1 | 5 | 1179 | 53 | ||
NM_001324227.1 | NP_001311156.1 | 6 | 877 | 53 | ||
NM_001324228.1 | NP_001311157.1 | 6 | 941 | 53 | ||
NM_001324229.1 | NP_001311158.1 | 5 | 1368 | 53 | ||
NM_001324230.1 | NP_001311159.1 | 7 | 1194 | 53 | ||
NM_004809.4 | NP_004800.2 | 6 | 1063 | 53 | ||
XM_005254792.4 | XP_005254849.1 | 6 | 1967 | 53 | ||
XM_006720773.3 | XP_006720836.1 | 6 | 1965 | 53 | ||
GenBank mRNA | AF074953.1 | - | 6 | 925 | 53 | |
AF111800.1 | - | 5 | 862 | 53 | ||
AF156557.1 | - | 6 | 1071 | 53 | ||
AK075244.1 | - | 6 | 959 | 53 | ||
AK093689.1 | - | 2 | 1328 | 53 | ||
AK222665.1 | - | 6 | 943 | 53 | ||
AK300799.1 | - | 7 | 1075 | 53 | ||
AK302051.1 | - | 5 | 868 | 53 | ||
AL109664.1 | - | 5 | 740 | 53 | ||
AL109665.1 | - | 6 | 914 | 53 | ||
AL533460.3 | - | 6 | 851 | 53 | ||
BC034379.1 | - | 6 | 1009 | 5185908 | 53 | |
BC065249.1 | - | 6 | 1009 | 6059393 | 53 | |
CR542089.1 | - | 6 | 938 | 53 | ||
Y16522.1 | - | 6 | 970 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |