Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029232.hg.1 |
ENST00000392665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029232.hg.1 |
ENST00000392666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029232.hg.1 |
NM_000220.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029232.hg.1 |
NM_153764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029232.hg.1 |
NM_153765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029232.hg.1 |
NM_153766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029232.hg.1 |
NM_153767.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029232.hg.1 |
TC11002443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029235.hg.1 |
ENST00000324036.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029235.hg.1 |
ENST00000392664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029235.hg.1 |
ENST00000392665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029235.hg.1 |
ENST00000392666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029235.hg.1 |
ENST00000440599.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029235.hg.1 |
NM_000220.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029235.hg.1 |
NM_153764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029235.hg.1 |
NM_153765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029235.hg.1 |
NM_153766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029235.hg.1 |
NM_153767.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029235.hg.1 |
TC11002443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029233.hg.1 |
ENST00000392665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029233.hg.1 |
ENST00000392666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029233.hg.1 |
ENST00000440599.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029233.hg.1 |
NM_000220.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029233.hg.1 |
NM_153764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029233.hg.1 |
NM_153765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029233.hg.1 |
NM_153766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029233.hg.1 |
NM_153767.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029233.hg.1 |
TC11002443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029234.hg.1 |
ENST00000324036.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029234.hg.1 |
ENST00000392665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029234.hg.1 |
ENST00000392666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029234.hg.1 |
ENST00000440599.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029234.hg.1 |
NM_000220.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029234.hg.1 |
NM_153764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029234.hg.1 |
NM_153765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029234.hg.1 |
NM_153766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029234.hg.1 |
NM_153767.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029234.hg.1 |
TC11002443.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029237.hg.1 |
ENST00000324036.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029237.hg.1 |
ENST00000392664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029237.hg.1 |
ENST00000392665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029237.hg.1 |
ENST00000392666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029237.hg.1 |
ENST00000440599.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029237.hg.1 |
NM_000220.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029237.hg.1 |
NM_153764.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029237.hg.1 |
NM_153765.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029237.hg.1 |
NM_153766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029237.hg.1 |
NM_153767.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11029237.hg.1 |
TC11002443.hg.1 |
HTA TC |