Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000543.4 | NP_000534.3 | 3 - 4 | 1441 | 104 | |
NM_001007593.2 | NP_001007594.2 | 3 - 4 | 1438 | 104 | ||
NM_001318087.1 | NP_001305016.1 | 3 - 4 | 1441 | 104 | ||
NM_001318088.1 | NP_001305017.1 | 3 - 4 | 1481 | 104 | ||
NR_134502.1 | 3 - 4 | 788 | 104 | |||
GenBank mRNA | AB209775.1 | - | 3 - 4 | 1424 | 104 | |
AK292388.1 | - | 3 - 4 | 1372 | 104 | ||
AY649987.1 | - | 3 - 4 | 620 | 104 | ||
HQ132745.1 | - | 3 - 4 | 1250 | 104 | ||
HQ132746.1 | - | 3 - 4 | 1181 | 104 | ||
HQ132747.1 | - | 3 - 4 | 1250 | 104 | ||
M59916.1 | - | 3 - 4 | 1350 | 104 | ||
X52678.1 | - | 2 - 3 | 869 | 104 | ||
X59960.1 | - | 3 - 4 | 1366 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |