Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009697.hg.1 |
TC07000632.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009697.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07009697.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008563.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160377.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160377.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160377.hg.1 |
ENST00000252723.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160377.hg.1 |
NM_000799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160377.hg.1 |
TC0700008563.hg.1 |
Clariom D TC |