Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR5H14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005514.1 | NP_001005514.1 | 1 | 243 | 111 | |
GenBank mRNA | BC136984.1 | - | 1 | 303 | 9020976 | 111 |
Set Membership: |
Detect Genomic DNA Both primers and probe map within a single exon |