Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008501.hg.1 |
ENST00000426354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008501.hg.1 |
ENST00000426354.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011972.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011972.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011972.hg.1 |
TC22000675.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011974.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011974.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011974.hg.1 |
TC22000675.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000490640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000490640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000490640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
ENST00000490640.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011973.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011973.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011973.hg.1 |
TC22000675.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000490640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000490640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000490640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
ENST00000490640.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008500.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008501.hg.1 |
ENST00000426354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008501.hg.1 |
ENST00000426354.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161217.hg.1 |
TC2200008500.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200161217.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161217.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161217.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161217.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161217.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161217.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161219.hg.1 |
TC2200008500.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200161219.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161219.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161219.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161219.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161219.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161219.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161216.hg.1 |
TC2200008500.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200161216.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161216.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161216.hg.1 |
NM_001010859.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200161216.hg.1 |
ENST00000382097.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161216.hg.1 |
ENST00000467813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200161216.hg.1 |
NM_001010859.2 |
Refseq |