Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033055.2 | NP_001028227.1 | 2 - 3 | 386 | 102 | |
NM_001285399.2 | NP_001272328.1 | 3 - 4 | 530 | 102 | ||
NM_001285400.2 | NP_001272329.1 | 3 - 4 | 500 | 102 | ||
NM_001285401.2 | NP_001272330.1 | 3 - 4 | 566 | 102 | ||
NM_001285402.1 | NP_001272331.1 | 3 - 4 | 557 | 102 | ||
NM_014336.4 | NP_055151.3 | 3 - 4 | 566 | 102 | ||
GenBank mRNA | AB593043.1 | - | 3 - 4 | 487 | 102 | |
AB593044.1 | - | 3 - 4 | 501 | 102 | ||
AB593045.1 | - | 3 - 4 | 501 | 102 | ||
AB593046.1 | - | 3 - 4 | 503 | 102 | ||
AB593047.1 | - | 3 - 4 | 544 | 102 | ||
AB593048.1 | - | 3 - 4 | 557 | 102 | ||
AB593049.1 | - | 3 - 4 | 511 | 102 | ||
AB593050.1 | - | 3 - 4 | 487 | 102 | ||
AB593051.1 | - | 3 - 4 | 488 | 102 | ||
AB593052.1 | - | 3 - 4 | 487 | 102 | ||
AB593055.1 | - | 3 - 4 | 532 | 102 | ||
AF038437.1 | - | 3 - 4 | 485 | 102 | ||
AF148864.1 | - | 3 - 4 | 651 | 102 | ||
AJ633677.1 | - | 2 - 3 | 339 | 102 | ||
AJ830742.1 | - | 3 - 4 | 454 | 102 | ||
AJ830743.1 | - | 3 - 4 | 520 | 102 | ||
AK023970.1 | - | 3 - 4 | 494 | 102 | ||
AK314558.1 | - | 3 - 4 | 487 | 102 | ||
BC012055.1 | - | 3 - 4 | 479 | 4634156 | 102 | |
BX537907.1 | - | 3 - 4 | 464 | 102 | ||
BX647279.1 | - | 3 - 4 | 532 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |