Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001314.hg.1 |
ENST00000339180.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001314.hg.1 |
ENST00000342473.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001314.hg.1 |
ENST00000359320.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001314.hg.1 |
ENST00000382809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001314.hg.1 |
ENST00000397227.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001314.hg.1 |
ENST00000555731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001314.hg.1 |
NM_001037288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001314.hg.1 |
NM_138460.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001314.hg.1 |
TC14000146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001311.hg.1 |
ENST00000339180.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001311.hg.1 |
ENST00000359320.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001311.hg.1 |
ENST00000382809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001311.hg.1 |
ENST00000555731.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001311.hg.1 |
NM_001037288.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001311.hg.1 |
NM_138460.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14001311.hg.1 |
TC14000146.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000339180.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000359320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000382809.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000553750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000555487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000555731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
NM_001037288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
NM_001288746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
NM_138460.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000339180.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000359320.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000382809.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000553750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000555487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
ENST00000555731.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
NM_001037288.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
NM_001288746.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
NM_138460.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400145796.hg.1 |
TC1400006686.hg.1 |
Clariom D TC |