Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033159.3 | NP_149349.2 | 2 - 3 | 1063 | 80 | |
NM_153281.1 | NP_695013.1 | 4 - 5 | 1330 | 80 | ||
NM_153283.2 | NP_695015.1 | 2 - 3 | 706 | 80 | ||
NM_153285.2 | NP_695017.1 | 2 - 3 | 288 | 80 | ||
NR_047690.1 | 1 - 2 | 1548 | 80 | |||
XM_011533667.2 | XP_011531969.1 | 3 - 4 | 1453 | 80 | ||
XM_011533668.2 | XP_011531970.1 | 2 - 3 | 1344 | 80 | ||
XM_011533669.2 | XP_011531971.1 | 3 - 4 | 1741 | 80 | ||
GenBank mRNA | AF173154.1 | - | 2 - 3 | 1034 | 80 | |
AF502905.1 | - | 2 - 3 | 678 | 80 | ||
AF502907.1 | - | 2 - 3 | 260 | 80 | ||
AK097287.1 | - | 4 - 5 | 1330 | 80 | ||
AK290816.1 | - | 2 - 3 | 1055 | 80 | ||
BC025774.1 | - | 2 - 3 | 1011 | 80 | ||
BC035695.1 | - | 2 - 3 | 1017 | 5760636 | 80 | |
KU178142.1 | - | 1 - 2 | 903 | 80 | ||
U03056.2 | - | 1 - 2 | 1519 | 80 | ||
U96078.1 | - | 1 - 2 | 1513 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons |