Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ012833.1 | - | 1 - 2 | 237 | 113 | |
AJ012834.1 | - | 1 - 2 | 321 | 113 | ||
AJ012835.1 | - | 1 - 2 | 323 | 113 | ||
AJ223040.1 | - | 1 - 2 | 332 | 113 | ||
AJ223041.1 | - | 1 - 2 | 320 | 113 | ||
AY679088.1 | - | 1 - 2 | 320 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |