Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019013.hg.1 |
TC01001234.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019013.hg.1 |
ENST00000335535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019013.hg.1 |
ENST00000368777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01019013.hg.1 |
NM_178356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010002.hg.1 |
ENST00000335535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010002.hg.1 |
ENST00000335535.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010002.hg.1 |
ENST00000368777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010002.hg.1 |
ENST00000368777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010002.hg.1 |
ENST00000368777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010002.hg.1 |
ENST00000368777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010002.hg.1 |
NM_178356.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010002.hg.1 |
NM_178356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172503.hg.1 |
ENST00000335535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172503.hg.1 |
ENST00000368777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172503.hg.1 |
NM_178356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172503.hg.1 |
ENST00000335535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172503.hg.1 |
ENST00000368777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100172503.hg.1 |
NM_178356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100172503.hg.1 |
TC0100010002.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010002.hg.1 |
ENST00000335535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010002.hg.1 |
ENST00000335535.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010002.hg.1 |
ENST00000368777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010002.hg.1 |
ENST00000368777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010002.hg.1 |
ENST00000368777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010002.hg.1 |
ENST00000368777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010002.hg.1 |
NM_178356.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010002.hg.1 |
NM_178356.3 |
Refseq |