Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015860.hg.1 |
TC03000908.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015860.hg.1 |
ENST00000335556.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015860.hg.1 |
ENST00000428432.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03015860.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167894.hg.1 |
TC0300009455.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009455.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
ENST00000335556.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
ENST00000428432.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
ENST00000487910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
NM_001304366.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
NM_182610.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
NR_130713.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300167893.hg.1 |
TC0300009455.hg.1 |
Clariom D TC |