Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001508.2 | NP_001499.1 | 1 - 2 | 1330 | 58 | |
GenBank mRNA | AF034633.1 | - | 1 - 2 | 861 | 58 | |
AK122643.1 | - | 1 - 2 | 1330 | 58 | ||
AK314895.1 | - | 1 - 2 | 1087 | 58 | ||
BC125046.1 | - | 1 - 2 | 861 | 40116106 | 58 | |
BC125047.1 | - | 1 - 2 | 850 | 58 | ||
FJ348258.1 | - | 1 - 2 | 861 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |