Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008140.hg.1 |
ENST00000338862.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008140.hg.1 |
ENST00000377942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008140.hg.1 |
ENST00000418407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008140.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008140.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008140.hg.1 |
TC13000693.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000613548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000613548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000613548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
ENST00000613548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008139.hg.1 |
ENST00000338862.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008139.hg.1 |
ENST00000377942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008139.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008139.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13008139.hg.1 |
TC13000693.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300161080.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161080.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161080.hg.1 |
ENST00000613548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161080.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161080.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161080.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161080.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161080.hg.1 |
ENST00000613548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161080.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161080.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161080.hg.1 |
TC1300009026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300161079.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161079.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161079.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161079.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161079.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161079.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300161079.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161079.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300161079.hg.1 |
TC1300009026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000338862.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000377942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000613548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000613548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000613548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
ENST00000613548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_002590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009026.hg.1 |
NM_032949.2 |
Refseq |