Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003700.1 | NP_003691.1 | 1 | 740 | 101 | |
GenBank mRNA | BC140729.1 | - | 1 | 839 | 9021597 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |